Assay Details
| Gene Symbol: cryz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005689.1 | NP_001005689.1 | 3 - 4 | 315 | 72 | |
| XM_012960813.2 | XP_012816267.1 | 3 - 4 | 316 | 72 | ||
| XM_012960814.2 | XP_012816268.1 | 3 - 4 | 295 | 72 | ||
| GenBank mRNA | BC075114.1 | - | 3 - 4 | 315 | 72 | |
More Information
Set Membership: |
|

cryz
Genomic Map