Assay Details
| Gene Symbol: sod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005694.1 | NP_001005694.1 | 1 - 2 | 72 | 55 | |
| GenBank mRNA | BC075257.1 | - | 1 - 2 | 72 | 55 | |
| CR761857.2 | - | 1 - 2 | 122 | 55 | ||
More Information
Set Membership: |
|

sod2
Genomic Map