Assay Details
| Gene Symbol: fcn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006717.2 | NP_001006718.1 | 3 - 4 | 303 | 86 | |
| XM_012969759.2 | XP_012825213.1 | 2 - 3 | 249 | 86 | ||
| GenBank mRNA | BC075449.1 | - | 3 - 4 | 305 | 86 | |
More Information
Set Membership: |
|

fcn3
Genomic Map