Assay Details
| Gene Symbol: rnf126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006734.1 | NP_001006735.1 | 1 - 2 | 193 | 78 | |
| XM_012959798.2 | XP_012815252.1 | 1 - 2 | 206 | 78 | ||
| GenBank mRNA | BC075492.1 | - | 1 - 2 | 193 | 78 | |
| CR848559.2 | - | 1 - 2 | 191 | 78 | ||
More Information
Set Membership: |
|

rnf126
Genomic Map