Assay Details
| Gene Symbol: tubd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006746.1 | NP_001006747.1 | 5 - 6 | 911 | 73 | |
| GenBank mRNA | BC075514.1 | - | 5 - 6 | 911 | 73 | |
More Information
Set Membership: |
|

tubd1
Genomic Map