Assay Details
| Gene Symbol: wnt5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006766.1 | NP_001006767.1 | 3 - 4 | 467 | 65 | |
| XM_012958845.2 | XP_012814299.1 | 3 - 4 | 941 | 65 | ||
| GenBank mRNA | BC075560.1 | - | 3 - 4 | 467 | 65 | |
| CR760740.2 | - | 2 - 3 | 357 | 65 | ||
More Information
Set Membership: |
|

wnt5b
Genomic Map