Assay Details
| Gene Symbol: rab34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006768.1 | NP_001006769.1 | 2 - 3 | 268 | 63 | |
| GenBank mRNA | BC075563.1 | - | 2 - 3 | 268 | 63 | |
More Information
Set Membership: |
|

rab34
Genomic Map