Assay Details
| Gene Symbol: apex2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006803.1 | NP_001006804.1 | 4 - 5 | 491 | 66 | |
| GenBank mRNA | BC076676.1 | - | 4 - 5 | 491 | 66 | |
| CR762042.2 | - | 4 - 5 | 579 | 66 | ||
| CU075608.2 | - | 4 - 5 | 502 | 66 | ||
More Information
Set Membership: |
|

apex2
Genomic Map