Assay Details
| Gene Symbol: apex2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006803.1 | NP_001006804.1 | 5 - 6 | 634 | 64 | |
| GenBank mRNA | BC076676.1 | - | 5 - 6 | 634 | 64 | |
| CR762042.2 | - | 5 - 6 | 722 | 64 | ||
| CU075608.2 | - | 5 - 6 | 645 | 64 | ||
More Information
Set Membership: |
|

apex2
Genomic Map