Assay Details
| Gene Symbol: apex2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006803.1 | NP_001006804.1 | 6 - 7 | 705 | 63 | |
| GenBank mRNA | BC076676.1 | - | 6 - 7 | 705 | 63 | |
| CR762042.2 | - | 6 - 7 | 793 | 63 | ||
| CU075608.2 | - | 6 - 7 | 716 | 63 | ||
More Information
Set Membership: |
|

apex2
Genomic Map