Assay Details
| Gene Symbol: calcoco2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006827.1 | NP_001006828.1 | 13 - 14 | 1379 | 67 | |
| GenBank mRNA | BC076895.1 | - | 13 - 14 | 1379 | 67 | |
| CR926335.2 | - | 13 - 14 | 1422 | 67 | ||
More Information
Set Membership: |
|

calcoco2
Genomic Map