Assay Details
| Gene Symbol: dtymk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006833.1 | NP_001006834.1 | 1 - 2 | 284 | 84 | |
| GenBank mRNA | BC076907.1 | - | 1 - 2 | 284 | 84 | |
| CR761321.2 | - | 1 - 2 | 229 | 84 | ||
More Information
Set Membership: |
|

dtymk
Genomic Map