Assay Details
| Gene Symbol: prtn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006846.1 | NP_001006847.1 | 1 - 2 | 61 | 82 | |
| GenBank mRNA | BC076933.1 | - | 1 - 2 | 61 | 82 | |
More Information
Set Membership: |
|

prtn3
Genomic Map