Assay Details
| Gene Symbol: cct4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006851.1 | NP_001006852.1 | 11 - 12 | 1357 | 70 | |
| XM_012962893.2 | XP_012818347.1 | 11 - 12 | 1578 | 70 | ||
| GenBank mRNA | BC076940.1 | - | 11 - 12 | 1357 | 70 | |
| CR761022.2 | - | 11 - 12 | 1399 | 70 | ||
More Information
Set Membership: |
|

cct4
Genomic Map