Assay Details
| Gene Symbol: znf330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006859.1 | NP_001006860.1 | 5 - 6 | 423 | 62 | |
| XM_012960091.2 | XP_012815545.1 | 5 - 6 | 448 | 62 | ||
| XM_012960095.2 | XP_012815549.1 | 5 - 6 | 462 | 62 | ||
| XM_012960097.1 | XP_012815551.1 | 5 - 6 | 413 | 62 | ||
| GenBank mRNA | BC076967.1 | - | 5 - 6 | 423 | 62 | |
| CR848239.2 | - | 5 - 6 | 328 | 62 | ||
More Information
Set Membership: |
|

znf330
Genomic Map