Assay Details
| Gene Symbol: phf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006860.1 | NP_001006861.1 | 1 - 2 | 486 | 67 | |
| XM_012966441.2 | XP_012821895.1 | 1 - 2 | 674 | 67 | ||
| GenBank mRNA | BC076968.1 | - | 1 - 2 | 486 | 67 | |
| CT030461.1 | - | 1 - 2 | 467 | 67 | ||
More Information
Set Membership: |
|

phf13
Genomic Map