Assay Details
| Gene Symbol: phf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006860.1 | NP_001006861.1 | 3 - 4 | 1198 | 79 | |
| XM_012966441.2 | XP_012821895.1 | 3 - 4 | 1386 | 79 | ||
| XM_012966442.2 | XP_012821896.1 | 3 - 4 | 851 | 79 | ||
| GenBank mRNA | BC076968.1 | - | 3 - 4 | 1198 | 79 | |
| CT030461.1 | - | 3 - 4 | 1179 | 79 | ||
More Information
Set Membership: |
|

phf13
Genomic Map