Assay Details
| Gene Symbol: rraga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006896.1 | NP_001006897.1 | 7 - 8 | 701 | 63 | |
| GenBank mRNA | BC074593.1 | - | 7 - 8 | 701 | 63 | |
More Information
Set Membership: |
|

rraga
Genomic Map