Assay Details
| Gene Symbol: rraga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006896.1 | NP_001006897.1 | 9 - 10 | 910 | 71 | |
| GenBank mRNA | BC074593.1 | - | 9 - 10 | 910 | 71 | |
| CR926416.2 | - | 8 - 9 | 873 | 71 | ||
More Information
Set Membership: |
|

rraga
Genomic Map