Assay Details
| Gene Symbol: srsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006918.1 | NP_001006919.1 | 2 - 3 | 693 | 66 | |
| GenBank mRNA | BC075558.1 | - | 2 - 3 | 693 | 66 | |
More Information
Set Membership: |
|

srsf1
Genomic Map