Assay Details
| Gene Symbol: h2afy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006924.1 | NP_001006925.1 | 5 - 6 | 903 | 87 | |
| GenBank mRNA | BC076893.1 | - | 5 - 6 | 903 | 87 | |
| CR942423.1 | - | 5 - 6 | 789 | 87 | ||
More Information
Set Membership: |
|

h2afy2
Genomic Map