Assay Details
| Gene Symbol: alas2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006925.1 | NP_001006926.1 | 3 - 4 | 388 | 63 | |
| XM_018096225.1 | XP_017951714.1 | 3 - 4 | 433 | 63 | ||
| GenBank mRNA | BC076921.1 | - | 3 - 4 | 388 | 63 | |
More Information
Set Membership: |
|

alas2
Genomic Map