Assay Details
| Gene Symbol: lpar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007474.1 | NP_001007475.1 | 2 - 3 | 876 | 97 | |
| XM_012953869.2 | XP_012809323.1 | 2 - 3 | 1100 | 97 | ||
| XM_012953870.2 | XP_012809324.1 | 2 - 3 | 919 | 97 | ||
| GenBank mRNA | AY652941.1 | - | 2 - 3 | 876 | 97 | |
| BC167261.1 | - | 2 - 3 | 862 | 97 | ||
| BC170629.1 | - | 2 - 3 | 779 | 97 | ||
| BC170631.1 | - | 2 - 3 | 779 | 97 | ||
| CT025445.2 | - | 2 - 3 | 860 | 97 | ||
More Information
Set Membership: |
|

lpar2
Genomic Map