Assay Details
| Gene Symbol: dstyk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007476.1 | NP_001007477.1 | 1 - 2 | 272 | 62 | |
| GenBank mRNA | AY364233.1 | - | 1 - 2 | 245 | 62 | |
| AY429679.1 | - | 1 - 2 | 272 | 62 | ||
| BC154679.1 | - | 1 - 2 | 260 | 62 | ||
| CR942484.2 | - | 1 - 2 | 256 | 62 | ||
More Information
Set Membership: |
|

dstyk
Genomic Map