Assay Details
| Gene Symbol: fam3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007502.1 | NP_001007503.1 | 8 - 9 | 611 | 66 | |
| XM_012960901.2 | XP_012816355.1 | 7 - 8 | 709 | 66 | ||
| XM_012960902.2 | XP_012816356.1 | 8 - 9 | 776 | 66 | ||
| XM_018089361.1 | XP_017944850.1 | 8 - 9 | 631 | 66 | ||
| GenBank mRNA | BC079939.1 | - | 8 - 9 | 611 | 66 | |
| CR760189.2 | - | 8 - 9 | 608 | 66 | ||
More Information
Set Membership: |
|

fam3d
Genomic Map