Assay Details
| Gene Symbol: arhgdig | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007515.1 | NP_001007516.1 | 4 - 5 | 429 | 90 | |
| XM_012970318.1 | XP_012825772.1 | 4 - 5 | 742 | 90 | ||
| GenBank mRNA | BC079956.1 | - | 4 - 5 | 429 | 90 | |
| Gene Symbol: act3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CT025518.2 | - | 370 | 90 | ||
More Information
Set Membership: |
|

arhgdig
Genomic Map