Assay Details
| Gene Symbol: rpl12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007859.1 | NP_001007860.1 | 3 | 519 | 69 | |
| GenBank mRNA | BC079961.1 | - | 3 | 519 | 69 | |
| CR761927.2 | - | 3 | 343 | 69 | ||
More Information
Set Membership: |
|

rpl12
Genomic Map