Assay Details
| Gene Symbol: meaf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007867.1 | NP_001007868.1 | 1 - 2 | 99 | 75 | |
| XM_012956794.2 | XP_012812248.1 | 1 - 2 | 185 | 75 | ||
| GenBank mRNA | BC080132.1 | - | 1 - 2 | 99 | 75 | |
More Information
Set Membership: |
|

meaf6
Genomic Map