Assay Details
| Gene Symbol: chga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007914.1 | NP_001007915.1 | 7 - 8 | 869 | 72 | |
| XM_012968262.2 | XP_012823716.1 | 6 - 7 | 965 | 72 | ||
| XM_012968263.2 | XP_012823717.1 | 7 - 8 | 910 | 72 | ||
| XM_012968264.2 | XP_012823718.1 | 6 - 7 | 965 | 72 | ||
| GenBank mRNA | BC080353.1 | - | 7 - 8 | 869 | 72 | |
| CR848134.2 | - | 6 - 7 | 849 | 72 | ||
More Information
Set Membership: |
|

chga
Genomic Map