Assay Details
| Gene Symbol: slc30a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007939.1 | NP_001007940.1 | 1 - 2 | 763 | 101 | |
| GenBank mRNA | BC080447.1 | - | 1 - 2 | 763 | 101 | |
More Information
Set Membership: |
|

slc30a1
Genomic Map