Assay Details
| Gene Symbol: aprt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007940.1 | NP_001007941.1 | 2 - 3 | 305 | 76 | |
| XM_018093014.1 | XP_017948503.1 | 2 - 3 | 173 | 76 | ||
| XM_018093015.1 | XP_017948504.1 | 2 - 3 | 374 | 76 | ||
| GenBank mRNA | BC080448.1 | - | 2 - 3 | 305 | 76 | |
| CR761604.2 | - | 2 - 3 | 316 | 76 | ||
| CT025478.2 | - | 2 - 3 | 314 | 76 | ||
More Information
Set Membership: |
|

aprt
Genomic Map