Assay Details
| Gene Symbol: qrich1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007959.1 | NP_001007960.1 | 9 - 10 | 2331 | 72 | |
| XM_012960927.2 | XP_012816381.1 | 9 - 10 | 2774 | 72 | ||
| XM_012960928.2 | XP_012816382.1 | 8 - 9 | 2666 | 72 | ||
| XM_012960929.2 | XP_012816383.1 | 8 - 9 | 2660 | 72 | ||
| GenBank mRNA | BC080472.1 | - | 9 - 10 | 2331 | 72 | |
More Information
Set Membership: |
|

qrich1
Genomic Map