Assay Details
| Gene Symbol: pdia6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007973.1 | NP_001007974.1 | 12 - 13 | 1345 | 75 | |
| GenBank mRNA | BC080483.1 | - | 12 - 13 | 1345 | 75 | |
| CR760891.2 | - | 12 - 13 | 1277 | 75 | ||
More Information
Set Membership: |
|

pdia6
Genomic Map