Assay Details
| Gene Symbol: psma4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007997.1 | NP_001007998.1 | 8 - 9 | 764 | 73 | |
| XM_018093016.1 | XP_017948505.1 | 8 - 9 | 818 | 73 | ||
| XM_018093017.1 | XP_017948506.1 | 8 - 9 | 850 | 73 | ||
| GenBank mRNA | BC080876.1 | - | 8 - 9 | 764 | 73 | |
| CR761624.2 | - | 8 - 9 | 739 | 73 | ||
More Information
Set Membership: |
|

psma4
Genomic Map