Assay Details
| Gene Symbol: cdc42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008026.2 | NP_001008027.1 | 5 - 6 | 561 | 100 | |
| GenBank mRNA | BC080906.1 | - | 5 - 6 | 561 | 100 | |
More Information
Set Membership: |
|

cdc42
Genomic Map