Assay Details
| Gene Symbol: rps3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008074.1 | NP_001008075.1 | 2 - 3 | 179 | 72 | |
| GenBank mRNA | BC080969.1 | - | 2 - 3 | 179 | 72 | |
| CR760859.2 | - | 2 - 3 | 176 | 72 | ||
More Information
Set Membership: |
|

rps3a
Genomic Map