Assay Details
| Gene Symbol: mob2.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008165.1 | NP_001008166.1 | 3 - 4 | 457 | 76 | |
| XM_018095358.1 | XP_017950847.1 | 4 - 5 | 758 | 76 | ||
| GenBank mRNA | BC082348.1 | - | 3 - 4 | 457 | 76 | |
More Information
Set Membership: |
|

mob2.2
Genomic Map