Assay Details
| Gene Symbol: ccz1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008197.2 | NP_001008198.1 | 10 - 11 | 858 | 76 | |
| XM_012970351.1 | XP_012825805.1 | 10 - 11 | 1397 | 76 | ||
| XM_018097092.1 | XP_017952581.1 | 10 - 11 | 850 | 76 | ||
| GenBank mRNA | BC082526.1 | - | 10 - 11 | 859 | 76 | |
| CR760737.2 | - | 10 - 11 | 841 | 76 | ||
More Information
Set Membership: |
|

ccz1b
Genomic Map