Assay Details
| Gene Symbol: crisp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008203.1 | NP_001008204.1 | 3 - 4 | 215 | 57 | |
| GenBank mRNA | BC080935.1 | - | 3 - 4 | 215 | 57 | |
| CT025286.2 | - | 3 - 4 | 204 | 57 | ||
More Information
Set Membership: |
|

crisp2
Genomic Map