Assay Details
| Gene Symbol: acp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008209.1 | NP_001008210.1 | 3 - 4 | 457 | 96 | |
| XM_012953156.2 | XP_012808610.1 | 3 - 4 | 433 | 96 | ||
| GenBank mRNA | BC081357.1 | - | 3 - 4 | 457 | 96 | |
| CR942420.2 | - | 3 - 4 | 430 | 96 | ||
More Information
Set Membership: |
|

acp5
Genomic Map