Assay Details
| Gene Symbol: casp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011068.1 | NP_001011068.1 | 2 - 3 | 159 | 60 | |
| GenBank mRNA | BC084448.1 | - | 2 - 3 | 159 | 60 | |
More Information
Set Membership: |
|

casp6
Genomic Map