Assay Details
| Gene Symbol: emp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011113.2 | NP_001011113.1 | 2 - 3 | 174 | 65 | |
| GenBank mRNA | BC084511.1 | - | 2 - 3 | 176 | 65 | |
More Information
Set Membership: |
|

emp3
Genomic Map