Assay Details
| Gene Symbol: med27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011132.1 | NP_001011132.1 | 3 - 4 | 515 | 77 | |
| XM_018096710.1 | XP_017952199.1 | 3 - 4 | 574 | 77 | ||
| GenBank mRNA | BC084171.1 | - | 3 - 4 | 515 | 77 | |
More Information
Set Membership: |
|

med27
Genomic Map