Assay Details
| Gene Symbol: csnk1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011137.1 | NP_001011137.1 | 2 - 3 | 118 | 72 | |
| GenBank mRNA | BC084453.1 | - | 2 - 3 | 118 | 72 | |
| CT025494.2 | - | 2 - 3 | 111 | 72 | ||
More Information
Set Membership: |
|

csnk1e
Genomic Map