Assay Details
| Gene Symbol: csnk1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011137.1 | NP_001011137.1 | 6 - 7 | 775 | 70 | |
| XM_012960985.2 | XP_012816439.1 | 6 - 7 | 881 | 70 | ||
| XM_012960986.2 | XP_012816440.1 | 6 - 7 | 883 | 70 | ||
| XM_012960987.2 | XP_012816441.1 | 5 - 6 | 744 | 70 | ||
| GenBank mRNA | BC084453.1 | - | 6 - 7 | 775 | 70 | |
| CT025494.2 | - | 6 - 7 | 768 | 70 | ||
More Information
Set Membership: |
|

csnk1e
Genomic Map