Assay Details
| Gene Symbol: rps16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011146.1 | NP_001011146.1 | 2 - 3 | 80 | 57 | |
| GenBank mRNA | BC084534.1 | - | 2 - 3 | 80 | 57 | |
| CR942750.2 | - | 1 - 2 | 50 | 57 | ||
More Information
Set Membership: |
|

rps16
Genomic Map