Assay Details
| Gene Symbol: cfl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011156.2 | NP_001011156.1 | 2 - 3 | 408 | 84 | |
| GenBank mRNA | BC084909.1 | - | 2 - 3 | 409 | 84 | |
More Information
Set Membership: |
|

cfl2
Genomic Map