Assay Details
| Gene Symbol: krt55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011224.1 | NP_001011224.1 | 6 - 7 | 1348 | 77 | |
| GenBank mRNA | BC087788.1 | - | 6 - 7 | 1348 | 77 | |
More Information
Set Membership: |
|

krt55
Genomic Map