Assay Details
| Gene Symbol: wisp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011237.1 | NP_001011237.1 | 3 - 4 | 580 | 57 | |
| GenBank mRNA | BC087808.1 | - | 3 - 4 | 580 | 57 | |
More Information
Set Membership: |
|

wisp2
Genomic Map