Assay Details
| Gene Symbol: lyplal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011253.1 | NP_001011253.1 | 199 | 85 | ||
| GenBank mRNA | BC086497.2 | - | 199 | 85 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lyplal1| Gene Symbol: lyplal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011253.1 | NP_001011253.1 | 199 | 85 | ||
| GenBank mRNA | BC086497.2 | - | 199 | 85 | ||
Set Membership: |
|